missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies ME0328
Description
- ME0328 is a cell-permeable and selective inhibitor of poly(ADP-ribose) polymerase 3 (PARP-3; IC50 = 0.89 μM; Lindgren et al.) PARP-3 is required for stabilization of the mitotic spindle and crucial for mitotic progression (Sharif-Askari et al.)
- Enhances vinorelbine-induced microtubule destabilization, leading to cell cycle arrest and apoptosis in breast cancer cells (Sharif-Askari et al.)
Specifications
Specifications
| CAS | 1445251-22-8 |
| Quantity | 1 mg |
| Molecular Formula | C19H19N3O2 |
| Purity | ≥98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?