missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10267-1-AP-20UL
This item is not returnable.
View return policy
Description
MEA1 gene ontology annotations related to this gene include spermatogenesis.Specifications
| MEA1 | |
| Polyclonal | |
| Unconjugated | |
| MEA1 | |
| HYS, male enhanced antigen 1, MEA, MEA 1, MEA1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17256, 4201, 685131 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q16626, Q5FVH7, Q64327 | |
| Mea1 | |
| MEA1 Fusion Protein Ag0375 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction