missing translation for 'onlineSavingsMsg'
Learn More

Mebhydrolin Napadisilate, Mikromol™

Product Code. 30050498
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050498 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050498 Supplier Mikromol Supplier No. MM1891.00
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Mebhydrolin Napadisilate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mebhydrolin Napadisilate
CAS 6153-33-9
Analyte or Component Names Mebhydrolin Napadisilate
Shipping Condition Room Temperature
Molecular Formula 2 C19 H20 N2 . C10 H8 O6 S2
Certifications/Compliance ISO 17034
Synonym 1,5-Naphthalenedisulfonic acid, compd. with 2,3,4,5-tetrahydro-2-methyl-5-(phenylmethyl)-1H-pyrido[4,3-b]indole (1:2), 1,5-Naphthalenedisulfonic acid, compd. with 5-benzyl-2,3,4,5-tetrahydro-2-methyl-1H-pyrido[4,3-b]indole (1:2) (8CI), 2H-Pyrido[4,3-b]indole, 5-benzyl-1,3,4,5-tetrahydro-2-methyl-, 1,5-naphthalenedisulfonate (2:1) (6CI), 1H-Pyrido[4,3-b]indole, 2,3,4,5-tetrahydro-2-methyl-5-(phenylmethyl)-, 1,5-naphthalenedisulfonate (2:1) (9CI), 1H-Pyrido[4,3-b]indole, 5-benzyl-2,3,4,5-tetrahydro-2-methyl-, 1,5-naphthalenedisulfonate (2:1) (8CI), Diazolin, Diazoline, Fabahistin, Incidal, Mebhydrolin napadisylate, Omeril
InChI Formula InChI=1S/2C19H20N2.C10H8O6S2/c2*1-20-12-11-19-17(14-20)16-9-5-6-10-18(16)21(19)13-15-7-3-2-4-8-15;11-17(12,13)9-5-1-3-7-8(9)4-2-6-10(7)18(14,15)16/h2*2-10H,11-14H2,1H3;1-6H,(H,11,12,13)(H,14,15,16)
SMILES CN1CCc2c(C1)c3ccccc3n2Cc4ccccc4.CN5CCc6c(C5)c7ccccc7n6Cc8ccccc8.OS(=O)(=O)c9cccc%10c(cccc9%10)S(=O)(=O)O
IUPAC Name 5-benzyl-2-methyl-3,4-dihydro-1H-pyrido[4,3-b]indole;naphthalene-1,5-disulfonic acid
Molecular Weight (g/mol) 841.05
Quantity 100 mg
Formula Weight 840.3015 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.