missing translation for 'onlineSavingsMsg'
Learn More

Mebhydrolin Napadisylate, TRC

Product Code. 30012422
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012422 100 g 100g
30010465 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012422 Supplier Toronto Research Chemicals Supplier No. TRCM201500100G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6153-33-9
Molecular Formula 2 C19 H20 N2 . C10 H8 O6 S2
Molecular Weight (g/mol) 841.05
Synonym 1,5-Naphthalenedisulfonic acid, compd. with 2,3,4,5-tetrahydro-2-methyl-5-(phenylmethyl)-1H-pyrido[4,3-b]indole (1:2), 1,5-Naphthalenedisulfonic acid, compd. with 5-benzyl-2,3,4,5-tetrahydro-2-methyl-1H-pyrido[4,3-b]indole (1:2) (8CI), 2H-Pyrido[4,3-b]indole, 5-benzyl-1,3,4,5-tetrahydro-2-methyl-, 1,5-naphthalenedisulfonate (2:1) (6CI), 1H-Pyrido[4,3-b]indole, 2,3,4,5-tetrahydro-2-methyl-5-(phenylmethyl)-, 1,5-naphthalenedisulfonate (2:1) (9CI), 1H-Pyrido[4,3-b]indole, 5-benzyl-2,3,4,5-tetrahydro-2-methyl-, 1,5-naphthalenedisulfonate (2:1) (8CI), Diazolin, Diazoline, Fabahistin, Incidal, Mebhydrolin napadisylate, Omeril
IUPAC Name 5-benzyl-2-methyl-3,4-dihydro-1H-pyrido[4,3-b]indole;naphthalene-1,5-disulfonic acid
SMILES CN1CCc2c(C1)c3ccccc3n2Cc4ccccc4.CN5CCc6c(C5)c7ccccc7n6Cc8ccccc8.OS(=O)(=O)c9cccc%10c(cccc9%10)S(=O)(=O)O

Specifications

Quantity 100 g
InChI Formula InChI=1S/2C19H20N2.C10H8O6S2/c2*1-20-12-11-19-17(14-20)16-9-5-6-10-18(16)21(19)13-15-7-3-2-4-8-15;11-17(12,13)9-5-1-3-7-8(9)4-2-6-10(7)18(14,15)16/h2*2-10H,11-14H2,1H3;1-6H,(H,11,12,13)(H,14,15,16)
Formula Weight 840.3015
Chemical Name or Material Mebhydrolin Napadisilate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.