missing translation for 'onlineSavingsMsg'
Learn More

Mebrofenin, Mikromol™

Product Code. 30052363
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052363 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30052363 Supplier Mikromol Supplier No. MM3594.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Mebrofenin
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mebrofenin
CAS 78266-06-5
Analyte or Component Names Mebrofenin
Shipping Condition Room Temperature
Molecular Formula C15 H19 Br N2 O5
Certifications/Compliance ISO/IEC 17025
Synonym N-[2-[(3-Bromo-2,4,6-trimethylphenyl)amino]-2-oxoethyl]-N-(carboxymethyl)glycine, 2: PN: US20040033243 PAGE: 23 claimed protein, Mebrofenin, SQ 26962
InChI Formula InChI=1S/C15H19BrN2O5/c1-8-4-9(2)15(10(3)14(8)16)17-11(19)5-18(6-12(20)21)7-13(22)23/h4H,5-7H2,1-3H3,(H,17,19)(H,20,21)(H,22,23)
SMILES Cc1cc(C)c(NC(=O)CN(CC(=O)O)CC(=O)O)c(C)c1Br
IUPAC Name 2-[[2-(3-bromo-2,4,6-trimethylanilino)-2-oxoethyl]-(carboxymethyl)amino]acetic acid
Molecular Weight (g/mol) 387.23
Quantity 100 mg
Formula Weight 386.0477 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.