missing translation for 'onlineSavingsMsg'
Learn More

Meclofenamic acid sodium salt, 99+%, Thermo Scientific™

Meclofenamic acid (meclofenamate, CAS # 644-62-2) is a nonsteroidal anti-inflammatory drug (NSAID) with antipyretic and antineoplastic activities.

Brand:  Thermo Scientific Alfa Aesar J60484.06

Quantity 5g

 View more versions of this product

Product Code. 15403259

  • ONLINE EXCLUSIVE PRICE
    155.00€
    139.65€ / 5g
    Save 15.35€
    9% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description

• Meclofenamic acid is an aminobenzoic acid-based NSAID that is characterized by two aryl moieties

• Meclofenamic acid is an inhibitor of arachidonate 5-lipoxygenase and prostaglandin-endoperoxide synthase

• Meclofenamic acid prevents pyrogen release and inhibits wound granulation; it may also act as a neoplastic

Application

• Meclofenamic acid can complex metals, making it of interest to drug delivery studies that target its toxic metal complexes to cancerous cell types

• It is suitable for laboratory research on pain, degeneration, wound healing, and tumor growth

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

6385-02-0
321.153
JKTYCTJLWZYQGM-UHFFFAOYSA-N
131673979
[HH].CC1=C(C(=C(C=C1)Cl)NC2=CC=CC=C2C(=O)O)Cl.[Na]
C14H13Cl2NNaO2
MFCD00077376
meclofenamate sodium
2-(2,6-dichloro-3-methylanilino)benzoic acid;molecular hydrogen;sodium

Specifications

6385-02-0
MFCD00077376
meclofenamate sodium
JKTYCTJLWZYQGM-UHFFFAOYSA-N
2-(2,6-dichloro-3-methylanilino)benzoic acid;molecular hydrogen;sodium
131673979
≥99%
C14H13Cl2NNaO2
5g
Soluble in water at 50mg/ml; Also soluble in DMF, DMSO or ethanol
[HH].CC1=C(C(=C(C=C1)Cl)NC2=CC=CC=C2C(=O)O)Cl.[Na]
321.153
318.13
Meclofenamic acid sodium salt
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.