Learn More
Meclofenamic acid sodium salt, 99+%, Thermo Scientific™
Meclofenamic acid (meclofenamate, CAS # 644-62-2) is a nonsteroidal anti-inflammatory drug (NSAID) with antipyretic and antineoplastic activities.
45.80€ - 139.65€
Chemical Identifiers
| CAS | 6385-02-0 |
|---|---|
| Molecular Formula | C14H13Cl2NNaO2 |
| Molecular Weight (g/mol) | 321.153 |
| MDL Number | MFCD00077376 |
| InChI Key | JKTYCTJLWZYQGM-UHFFFAOYSA-N |
| Synonym | meclofenamate sodium |
| PubChem CID | 131673979 |
| IUPAC Name | 2-(2,6-dichloro-3-methylanilino)benzoic acid;molecular hydrogen;sodium |
| SMILES | [HH].CC1=C(C(=C(C=C1)Cl)NC2=CC=CC=C2C(=O)O)Cl.[Na] |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15493249
|
Thermo Scientific Alfa Aesar
J60484.03 |
1g |
45.80€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15403259
|
Thermo Scientific Alfa Aesar
J60484.06 |
5g |
155.00€ 139.65€ / 5g Save 15.35€ 9% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Meclofenamic acid is an aminobenzoic acid-based NSAID that is characterized by two aryl moieties
• Meclofenamic acid is an inhibitor of arachidonate 5-lipoxygenase and prostaglandin-endoperoxide synthase
• Meclofenamic acid prevents pyrogen release and inhibits wound granulation; it may also act as a neoplastic
Application
• Meclofenamic acid can complex metals, making it of interest to drug delivery studies that target its toxic metal complexes to cancerous cell types
• It is suitable for laboratory research on pain, degeneration, wound healing, and tumor growth
Chemical Identifiers
| 6385-02-0 | |
| 321.153 | |
| JKTYCTJLWZYQGM-UHFFFAOYSA-N | |
| 131673979 | |
| [HH].CC1=C(C(=C(C=C1)Cl)NC2=CC=CC=C2C(=O)O)Cl.[Na] |
| C14H13Cl2NNaO2 | |
| MFCD00077376 | |
| meclofenamate sodium | |
| 2-(2,6-dichloro-3-methylanilino)benzoic acid;molecular hydrogen;sodium |
Specifications
| 6385-02-0 | |
| MFCD00077376 | |
| Soluble in water at 50mg/ml; Also soluble in DMF, DMSO or ethanol | |
| [HH].CC1=C(C(=C(C=C1)Cl)NC2=CC=CC=C2C(=O)O)Cl.[Na] | |
| 321.153 | |
| 318.13 | |
| Meclofenamic acid sodium salt |
| C14H13Cl2NNaO2 | |
| meclofenamate sodium | |
| JKTYCTJLWZYQGM-UHFFFAOYSA-N | |
| 2-(2,6-dichloro-3-methylanilino)benzoic acid;molecular hydrogen;sodium | |
| 131673979 | |
| ≥99% |
Safety and Handling
missing translation for 'einecsNumber' : 228-983-3
missing translation for 'rtecsNumber' : CB2975500
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.