missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MeCP2 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP3-05542-50ul
This item is not returnable.
View return policy
Description
MeCP2 Polyclonal antibody specifically detects MeCP2 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| MeCP2 | |
| Polyclonal | |
| Western Blot 1:10000-1:20000, Immunohistochemistry 1:1000-1:2000, Immunocytochemistry/ Immunofluorescence 1:1000-1:2000 | |
| AUTSX3, DKFZp686A24160, MeCp-2 protein, mental retardation, X-linked 79, methyl CpG binding protein 2 (Rett syndrome), X-linked 16 | |
| Full length human recombinant MeCP2 expressed in and purified from E. coli. | |
| 50 μg | |
| Cancer, Chromatin Research, Neuroscience | |
| 4204 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgY |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Supplied as an aliquot of IgY preparation | |
| Chicken | |
| IgY purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction