Learn More
Ansell MEDI-GRIP Latex Surgical Glove
Product Code. 17774949 Shop All Ansell Products
Change view
Click to view available options
Size:
5.5
6
6.5
7
7.5
8
8.5
9
Unit Size:
50 pairs
Product Code. Size unitSize
17774949 7.5 50 pairs
17734949 5.5 50 pairs
17744949 6 50 pairs
17754949 6.5 50 pairs
17764949 7 50 pairs
17784949 8 50 pairs
17794949 8.5 50 pairs
17704959 9 50 pairs
8 options
Product Code. 17774949
Brand: Ansell 330200775
Only null left
Add to basket
Edge
Add to basket
Latex, powder-free surgical gloves that provide everyday comfort, dexterity and reliable protection.
  • Proprietary anatomic-shaped glove formers to provide a comfortable fit and minimizes hand fatigue
  • Soft, malleable latex formulation with optimized thickness to provide good tactile sensitivity and dexterity
  • Polymer coated for fast and easy donning
Specifications
AQL 0.65
Color Natural
Disposable Single-use
Exterior Finish Fully Textured
Material Natural Rubber Latex
Powdered Powder Free
Size 7.5
Sterility Sterile