missing translation for 'onlineSavingsMsg'
Learn More

Medroxyprogesterone-17-acetate 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30315046
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30315046

Brand: Dr. Ehrenstorfer DREA14852015AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Medroxyprogesterone acetate
CAS 71-58-9
Analyte or Component Names Medroxyprogesterone acetate
Shipping Condition Room Temperature
Molecular Formula C24 H34 O4
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 6alpha-Methyl-3,20-dioxopregn-4-en-17-yl acetate, Medroxyprogesterone acetate, Megestrol Acetate Imp. A (EP), Amen, Aragest, Aragest 5, Asconale, CBP 1011, Clinofem, Clinovir, Colirest, Curretab, Cycrin, DMPA, Depo-Clinovir, Depo-Prodasone, Depo-Provera, Depo-Ralovera, Depo-SubQ Provera 104, Depo-progestin, Depo-promone, Depomedroxyprogesterone acetate, Depot-medroxyprogesterone acetate, Farlutal, Farlutin, G-Farlutal, Gestapuran, Hematrol, Hysron, Indivina, Lutopolar, Lutoral, MAP, MAP (steroid), MPA, MPA Gyn 5, Med-Pro, Medroprogesterone acetate, Medroxyprogesterone 17-acetate, Meprate, Metigestrona, NSC 21171, NSC 26386, Nadigest, Nidaxin, Novo-Medrone, Oragest, Perlutex, Perlutex Leo, Prempack C, Prodasone, Progespon, Progestalfa, Progeston, Progevera, Provera, Proverone, Ralovera, Repromap, Repromix, Sirprogen, Sodelut G, U 8839, Veramix, Pregn-4-ene-3,20-dione, 17-(acetyloxy)-6-methyl-, (6alpha)-, Pregn-4-ene-3,20-dione, 17-hydroxy-6alpha-methyl-, acetate (7CI,8CI), Progesterone, 17-hydroxy-6alpha-methyl-, acetate (6CI), (6alpha)-17-(Acetyloxy)-6-methylpregn-4-ene-3,20-dione, 17alpha-Acetoxy-6alpha-methylpregn-4-ene-3,20-dione, 17alpha-Acetoxy-6alpha-methylprogesterone, 17alpha-Hydroxy-6alpha-methyl-4-pregnene-3,20-dione 17-acetate, 17alpha-Hydroxy-6alpha-methylpregn-4-ene-3,20-dione acetate, 17alpha-Hydroxy-6alpha-methylprogesterone acetate, 6alpha-Methyl-17-acetoxyprogesterone, 6alpha-Methyl-17R-hydroxyprogesterone acetate, 6alpha-Methyl-17alpha-Hydroxy-4-pregnene-3,20-dione acetate, 6alpha-Methyl-17alpha-acetoxy-Delta4-pregnene-3,20-dione, 6alpha-Methyl-17alpha-acetoxypregn-4-ene-3,20-dione, 6alpha-Methyl-17alpha-hydroxyprogesterone acetate, Megestrol Acetate Impurity A
InChI Formula InChI=1S/C24H34O4/c1-14-12-18-19(22(4)9-6-17(27)13-21(14)22)7-10-23(5)20(18)8-11-24(23,15(2)25)28-16(3)26/h13-14,18-20H,6-12H2,1-5H3/t14-,18+,19-,20-,22+,23-,24-/m0/s1
SMILES C[C@H]1C[C@@H]2[C@H](CC[C@@]3(C)[C@H]2CC[C@]3(OC(=O)C)C(=O)C)[C@@]4(C)CCC(=O)C=C14
IUPAC Name [(6S,8R,9S,10R,13S,14S,17R)-17-acetyl-6,10,13-trimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 386.52
Quantity 1 mL
Formula Weight 386.2457 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.