Learn More
MEF2C Antibody (CL0368) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52925B
Product Code. 30301248
  • 737.00€ / 0.10mL
Description
MEF2C Monoclonal antibody specifically detects MEF2C in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MEF2C
Monoclonal
Biotin
DEL5q14.3, myocyte enhancer factor 2C, myocyte-specific enhancer factor 2C, polypeptide C
This antibody was developed using a recombinant protein derived from Q06413, with the exact immunogen sequence remaining proprietary.
0.1 mL
Neuroscience
4208
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence
CL0368
PBS
Mouse
Protein A purified