Learn More
Mefenamic acid, 98%
Description
Mefenamic acid has demonstrated antiproliferative activity against colorectal cancer cells. It has Analgesic, Anti-inflammatory activity. It is used to relieve mild to moderate pain, including headaches, dental pain, osteoarthritis and rheumatoid arthritis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Mefenamic acid has demonstrated antiproliferative activity against colorectal cancer cells. It has Analgesic, Anti-inflammatory activity. It is used to relieve mild to moderate pain, including headaches, dental pain, osteoarthritis and rheumatoid arthritis.
Solubility
It is soluble in acetone, chloroform, dichloromethane, methanol. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
Specifications
| Color | White |
| Quantity | 100 g |
| Merck Index | 14,5798 |
| Solubility Information | It is soluble in acetone,chloroform,dichloromethane,methanol. Insoluble in water. |
| Formula Weight | 241.29 |
| Percent Purity | 98% |
| Physical Form | Powder |
| Chemical Name or Material | Mefenamic acid |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.