Learn More
Micronova™ MegaClean™ Heavy-Duty Cleaner
BIOPHARMA
Product Code. 13137736 Shop All Micronova Products
Change view
Click to view available options
Volume (English):
1 gal.
5 gal.
Unit Size:
15.14 L
19 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (English) Unit Size
13137736 5 gal. 19 L
11932475 1 gal. 15.14 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13137736 Supplier Micronova™ Supplier No. MC15G
  • ONLINE EXCLUSIVE PRICE
    373.00€
    357.72€ / 19 L
    Save 15.28€
    4% Off
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A non-flammable, heavy duty cleaner and degreaser that removes grease, adhesives and photo-resist stains from most surfaces. Available in 1 or 5 gallon bottles and a 1 quart spray bottle.
The Micronova MegaClean™ Heavy-Duty Cleaner offers an effective alternative to toxic, flammable, solvent-based cleaners, such as acetone and isopropyl alcohol. Filtered to 0.1 microns, this cleaner is ideal for semiconductor, electronics, aerospace and clean-build operations.
  • Non-flammable, heavy duty cleanroom degreaser
  • Removes grease, adhesives and photo-resist stains from most surfaces
  • Ideal for semiconductor, electronics, aerospace and clean-build operations
  • Filtered to 0.1 microns
  • Available in 1 or 5 gallon refill bottles and a 1 quart spray bottle
  • Biodegradable
  • Water soluble
  • Effective at low concentrations
Specifications
Product Type MegaClean Cleaner
Color-coded No
Handle Type None
Sterility Non-sterile
Description Heavy Duty Detergent, Degreaser
For Use With (Application) Ideal spot cleaning solution and easily removes grease, adhesives and photo-resist stains from most surfaces, floors, high traffic areas, parts cleaning, adhesive removal
For Use With (Equipment) Mops and Buckets or Wipes and Sponge
Volume (English) 5 gal.