Learn More
Applied Biosystems™ Megaplex™ Primer Pools, Human Pools B v3.0
GREENER_CHOICE
Product Code. 11670089 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
50 reactions
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11670089 50 reactions 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11670089 Supplier Applied Biosystems™ Supplier No. 4444749
  • ONLINE EXCLUSIVE PRICE
    1420.00€
    1221.20€ / 1 sets
    Save 198.80€
    14% Off
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
Megaplex™ Primer Pools, Human Pools B v3.0
Megaplex™ Primer Pools, Human Pools B v3.0 is available under a single part number containing both the Megaplex™ RT Primers and Megaplex™ PreAmp Primers, Human Pools B v3.0. Megaplex™ RT Primers, Human Pool B v3.0 contains RT primers for 377 unique microRNAs and 4 controls. For use when assay sensitivity of the utmost importance, Megaplex™ PreAmp Primers significantly enhance the ability to detect low expressed microRNAs enabling the generation of a comprehensive expression profile using far less sample, as low as 1 ng of input total RNA. The microRNA targets represented in this pool tend be less well characterized, more narrowly expressed and⁄or expressed at a lower level compared to Human Pool A. Megaplex™ Primer Pools, Human Pools B v3.0 are intended for use with the TaqMan™ Array Human MicroRNA B Card v3.0, however, they can also be used to prepare cDNA for real-time analysis using the individual TaqMan™ MicroRNA Assays.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -20°C
GC-Rich PCR Performance Low
PCR Method qPCR
Primer RT Primers, PreAmp Primers
Product Line Megaplex
Product Type Primer Pool
Quantity 50 reactions
Species Human
Sufficient For 50 Reactions
For Use With (Application) miRNA analysis
No. of Reactions 50 Reactions