missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEIS1 Antibody (OTI1B4), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44993-30ul
This item is not returnable.
View return policy
Description
MEIS1 Monoclonal antibody specifically detects MEIS1 in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| MEIS1 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| 4211 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |
| Immunohistochemistry, Western Blot | |
| OTI1B4 | |
| Western Blot 1:2000, Immunohistochemistry 1:2000 | |
| homeobox protein Meis1, leukemogenic homolog protein, Meis homeobox 1, MGC43380, myeloid ecotropic viral integration site 1 homolog, myeloid ecotropic viral integration site 1 homolog (mouse) | |
| Human recombinant protein fragment corresponding to amino acids 176-390 of human MEIS1(NP_002389) produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction