missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanopsin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-74460-0.025mg
This item is not returnable.
View return policy
Description
Melanopsin Polyclonal specifically detects Melanopsin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Knockout Validated.
Specifications
| Melanopsin | |
| Polyclonal | |
| Unconjugated | |
| PBS, 1mg/ml BSA with 0.05% Sodium Azide | |
| OPN4 | |
| Synthetic peptide corresponding to residues E(455) Q K S K T P K T K R H L P S L D R R M(474) of rat melanopsin. | |
| 0.025 mg | |
| Vision | |
| 94233 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 0.1 - 0.2 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 5 μg/mL, Immunohistochemistry-Frozen 1:1000, Knockout Validated | |
| melanopsin, MGC142118, MOP, opsin 4 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human reactivity reported in scientific literature (PMID: 20971788). | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction