missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melatonin Receptor 1B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-14401
This item is not returnable.
View return policy
Description
Melatonin Receptor 1B Polyclonal antibody specifically detects Melatonin Receptor 1B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Melatonin Receptor 1B | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| FGQTL2, Mel 1b melatonin receptor, Mel1b receptor, MEL-1B-R, melatonin receptor 1B, melatonin receptor MEL1B, melatonin receptor type 1B, MT2 | |
| Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human Melatonin Receptor 1B. | |
| 50 μg | |
| GPCR, Neuroscience | |
| 4544 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction