missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melphalan, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Melphalan |
| CAS | 148-82-3 |
| Molecular Formula | C13 H18 Cl2 N2 O2 |
| Quantity | 5 mg |
| Synonym | Melphalan, 4-[Bis(2-chloroethyl)amino]-l-phenylalanine, L-Phenylalanine, 4-[bis(2-chloroethyl)amino]-, Alanine, 3-[p-[bis(2-chloroethyl)amino]phenyl]-, L- (8CI), 3025CB, 4-Bis(2-chloroethyl)amino-L-phenylalanine, 67: PN: WO2010011890 PAGE: 37 claimed protein, Alanine nitrogen mustard, Alkeran, CB 3025, L-PAM, L-Phenylalanine mustard, L-Phenylalanine mustard hydrochloride, L-Sarcolysin, L-Sarcolysine, L-Sarkolysin, Levofalan, Levofolan, Levopholan, Melfalan, Melphalan, NSC 241286, NSC 8806, Phenylalanine mustard, Sarcoclorin |
| InChI Formula | InChI=1S/C13H18Cl2N2O2/c14-5-7-17(8-6-15)11-3-1-10(2-4-11)9-12(16)13(18)19/h1-4,12H,5-9,16H2,(H,18,19)/t12-/m0/s1 |
| SMILES | N[C@@H](Cc1ccc(cc1)N(CCCl)CCCl)C(=O)O |
| IUPAC Name | (2S)-2-amino-3-[4-[bis(2-chloroethyl)amino]phenyl]propanoic acid |
| Molecular Weight (g/mol) | 305.2 |
| Formula Weight | 304.0745 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction