missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Melrilimab Recombinant Monoclonal Antibody
GREENER_CHOICE

Recombinant Monoclonal Antibody

477.00€ - 1204.00€

Specifications

Antigen Melrilimab
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30284171
GREENER_CHOICE
View Documents
Invitrogen™
MA559553
100 μg
477.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30284101
View Documents
Invitrogen™
MA559554
1 mg
1204.00€
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Melrilimab is a biosimilar antibody, specifically an anti-IL1RL1/ST2 antibody. The gene associated with this biosimilar is IL1RL1, which encodes the interleukin-1 receptor-like 1 protein, also known as ST2 or IL-33R. This receptor is predominantly expressed on the surface of immune cells and plays a role in the immune response by interacting with the cytokine IL-33. IL1RL1 is located on chromosome 2q12.1 in humans. It is a member of the interleukin 1 receptor family and forms a heterodimer with the IL-1 receptor accessory protein (IL-1RAcP), functioning as the receptor for IL-33. IL1RL1 plays a crucial role in immune and inflammatory responses. It is particularly important in conditions such as asthma and rheumatoid arthritis, where it is upregulated. The protein structure includes an extracellular domain for ligand binding, a transmembrane domain, and an intracellular domain for signal transduction. IL-33 binding to IL1RL1 activates signaling pathways that lead to the production of pro-inflammatory cytokines and chemokines, contributing to the amplification of immune responses.
Specifications

Specifications

Melrilimab
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG2SA
Human
Primary
Protein A
1 mg/mL
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Unconjugated
Human
RUO
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.