missing translation for 'onlineSavingsMsg'
Learn More

Memantine hydrochloride, Thermo Scientific™

 

Brand:  Thermo Scientific Alfa Aesar J63830.MF


Quantity 50mg

Product Code. 15475989

  • 145.00€ / 50mg
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

41100-52-1
MFCD00214336
14,5829
LDDHMLJTFXJGPI-UHFFFAOYNA-N
hydrogen 3,5-dimethyladamantan-1-amine chloride
181458
215.76
C12H22ClN
50mg
memantine hydrochloride, memantine hcl, 3,5-dimethyladamantan-1-amine hydrochloride, 3,5-dimethyl-1-adamantanamine hydrochloride, namenda, akatinol, axura, namenda xr, memantine.hcl
[H+].[Cl-].CC12CC3CC(C)(C1)CC(N)(C3)C2
215.77
CHEBI:64323
Memantine hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Memantine hydrochloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contain: Memantine hydrochloride

SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.