Learn More
Veolia Water Systems Reverse Osmosis Membrane
Description
- RO membranes remove water contaminants that are less than 1 nm diameter
- They typically remove over 99% of ionic contamination, most organic contamination, and nearly all particulate contamination.
- Compatible with BIOPURE 7/15, MEDICA-D 7/15, MEDICA-R 7/15, MEDICA-S 7/15, PURELAB Option-E 5/10, PURELAB Option-Q, PURELAB Option-R 7/15, PURELAB Option-S 7/15, PURELAB Prima 7/15/30, PURELAB Pulse 1/2
Specifications
Specifications
| Flow Rate | 7.5 L/hr. |
| For Use With (Application) | Removing water contaminants that are less than 1 nm in diameter |
| For Use With (Equipment) | BIOPURE 7/15, MEDICA-D 7/15, MEDICA-R 7/15, MEDICA-S 7/15, PURELAB Option-E 5/10, PURELAB Option-Q, PURELAB Option-R 7/15, PURELAB Option-S 7/15, PURELAB Prima 7/15/30, PURELAB Pulse 1/2 |
| Type | Reverse Osmosis Membrane |