Learn More
Gilac™ Men's Urinal with Lid
Product Code. 17948551
Change view
Click to view available options
Volume (Metric):
1 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
17948551 1 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17948551 Supplier Gilac™ Supplier No. G101708
  • ONLINE EXCLUSIVE PRICE
    7.94€
    5.20€ / Pack of 1
    Save 2.74€
    34% Off
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Men's 1 L Urinal with Lid is graduated, and suitable for hospital use.
  • Suitable for hospital use.
  • Translucent, level visible, with volume graduations.
  • Rounded design and very thick.
  • Not easy to break when dropped.
  • Outstanding durability even in the intensive use cycle.
  • Product is disinfected in a bedpan washer.
  • Researched product ergonomics ensure optimum cleaning.
  • Equipment at room temperature to avoid the cold sensation experienced with stainless steel equivalents.
Specifications
Product Type Male Bed Bottle with Cap
Color Green
Material Polypropylene
Sterility Non-sterile
Graduated Yes
Volume (English) 0.3 gal.
Volume (Metric) 1 L