missing translation for 'onlineSavingsMsg'
Learn More

Menbutone, TRC

Product Code. 17901535
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17901535 1 g 1g
17912825 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17901535 Supplier Toronto Research Chemicals Supplier No. TRCM2187001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3562-99-0
Molecular Formula C15 H14 O4
Molecular Weight (g/mol) 258.27
Synonym 3-Butyl-4-hydroxyphenylacetic acid, Butylscopolamine
IUPAC Name 4-(4-methoxynaphthalen-1-yl)-4-oxobutanoic acid
SMILES COc1ccc(C(=O)CCC(=O)O)c2ccccc12

Specifications

Quantity 1 g
InChI Formula InChI=1S/C15H14O4/c1-19-14-8-6-11(13(16)7-9-15(17)18)10-4-2-3-5-12(10)14/h2-6,8H,7,9H2,1H3,(H,17,18)
Formula Weight 258.0892
Percent Purity >95%
Chemical Name or Material Menbutone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.