missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Menbutone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
87.20€ - 132.00€
Chemical Identifiers
| CAS | 3562-99-0 |
|---|---|
| Molecular Formula | C15 H14 O4 |
| Molecular Weight (g/mol) | 258.27 |
| Synonym | 3-Butyl-4-hydroxyphenylacetic acid, Butylscopolamine |
| IUPAC Name | 4-(4-methoxynaphthalen-1-yl)-4-oxobutanoic acid |
| SMILES | COc1ccc(C(=O)CCC(=O)O)c2ccccc12 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17912825
|
Toronto Research Chemicals
TRC-M218700-5G |
5 g |
132.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17901535
|
Toronto Research Chemicals
TRC-M218700-1G |
1 g |
87.20€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
| 3562-99-0 | |
| 258.27 | |
| 4-(4-methoxynaphthalen-1-yl)-4-oxobutanoic acid |
| C15 H14 O4 | |
| 3-Butyl-4-hydroxyphenylacetic acid, Butylscopolamine | |
| COc1ccc(C(=O)CCC(=O)O)c2ccccc12 |
Specifications
| 3562-99-0 | |
| 3-Butyl-4-hydroxyphenylacetic acid, Butylscopolamine | |
| COc1ccc(C(=O)CCC(=O)O)c2ccccc12 | |
| 258.27 | |
| >95% | |
| HPLC |
| C15 H14 O4 | |
| InChI=1S/C15H14O4/c1-19-14-8-6-11(13(16)7-9-15(17)18)10-4-2-3-5-12(10)14/h2-6,8H,7,9H2,1H3,(H,17,18) | |
| 4-(4-methoxynaphthalen-1-yl)-4-oxobutanoic acid | |
| 258.0892 | |
| Menbutone |
Safety and Handling
Recommended Storage : -20°C
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.