missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEPE Monoclonal Antibody (341611)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561269
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Matrix extracellular phosphoglycoprotein (MEPE), also known as osteoblast/osteocyte factor 45 (OF45), is a member of the small integrin-binding ligand N-linked glycoprotein (SIBLING) family. MEPE promotes renal phosphate excretion and modulates mineralization. It can be phosphorylated by secreted protein kinases. MEPE is expressed in osteoblasts, osteocytes and odontoblasts, and the expression has also been reported in numerous human tumor cell lines. It is a 525-amino acid protein with a calculated molecular weight of 58 kDa.
Specifications
| MEPE | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9NQ76 | |
| MEPE | |
| Mouse myeloma cell line NS0-derived recombinant human MEPE/OF45, Pro18-Asp525. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 341611 | |
| Unconjugated | |
| MEPE | |
| Matrix extracellular phosphoglycoprotein; matrix extracellular phosphoglycoprotein with ASARM motif (bone); matrix, extracellular phosphoglycoprotein with ASARM motif (bone); Mepe; OF45; osteoblast/osteocyte factor 45; Osteoregulin | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 56955 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction