Learn More
Hanna Instruments™ Mercury Free COD Test Tubes
Product Code. 11702435
Change view
Click to view available options
Volume (Metric):
0 to 15,000 mg/L
0 to 150 mg/L
Unit Size:
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11702435 0 to 150 mg/L Pack of 25
11722435 0 to 15,000 mg/L Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11702435 Supplier Hanna Instruments™ Supplier No. HI93754D25
  • ONLINE EXCLUSIVE PRICE
    160.65€
    85.10€ / Pack of 25
    Save 75.55€
    47% Off
 Restricted Item
Estimated Shipment: 26-08-2026
Specifications
Description Mercury Free COD Test Tube
Volume (Metric) 0 to 150 mg/L
For Use With (Application) For COD Determination
Quantity 25 Pack
compliance-icons compliance-icons
Product Identifier
  • X25 TUBE TEST COD 0-150MG/L WH HG
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P391-Collect spillage.
Supplemental information
  • MIXTURE LIST-Contain: sulphuric acid, silver sulphate, potassium dichromate