missing translation for 'onlineSavingsMsg'
Learn More

Merriam Crater Basalt, CRM, ARMI∣MBH

Product Code. 30040968
Change view
Click to view available options
Quantity:
75 g
Unit Size:
75g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040968 75 g 75g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30040968 Supplier ARMI/MBH Supplier No. MBHBASMC22P
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.

Merriam Crater Basalt, CRM
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfur/Lithium/Gadolinium/Arsenic/Tin/Beryllium/Germanium/Tungsten/Lutetium/Thulium/Cadmium/Antimony/Indium/Manganese/Manganese(II) oxide/Cesium/Phosphorus/Loss on ignition/Potassium/Phosphorus pentoxide/Potassium oxide/Terbium/Holmium/Titanium/Uranium/Molybdenum/Titanium dioxide/Ytterbium/Tantalum/Erbium/Europium/Sodium/Sodium oxide/Hafnium/Dysprosium/Magnesium/Lead/Thorium/Calcium/Samarium/Magnesium oxide/Iron/Aluminium/Rubidium/Calcium oxide/Praseodymium/Total Iron expressed as Fe2O3/Aluminium oxide/Gallium/Silicon/Yttrium/Scandium/Niobium/Silicon dioxide/Neodymium/Cobalt/Lanthanum/Copper/Chromium/Nickel/Zinc/Cerium/Zirconium/Vanadium/Barium/Strontium
CAS 14808-60-7
Quantity 75 g
Form Fine Powder
Concentration or Composition (by Analyte or Components) Sulfur (140) mg/kg/Lithium (7.6) mg/kg/Gadolinium (6.2) mg/kg/Arsenic (3.6) mg/kg/Tin (1.3) mg/kg/Beryllium (1.2) mg/kg/Germanium (0.6) mg/kg/Tungsten (0.32) mg/kg/Lutetium (0.31) mg/kg/Thulium (0.31) mg/kg/Cadmium (0.09) mg/kg/Antimony (0.08) mg/kg/Indium (0.075) mg/kg/Manganese 0.145 %/Manganese(II) oxide 0.187 %/Cesium 0.22 mg/kg/Phosphorus 0.315 %/Loss on ignition 0.6 %/Potassium 0.69 %/Phosphorus pentoxide 0.722 %/Potassium oxide 0.83 %/Terbium 0.85 mg/kg/Holmium 0.9 mg/kg/Titanium 1.12 %/Uranium 1.4 mg/kg/Molybdenum 1.6 mg/kg/Titanium dioxide 1.86 %/Ytterbium 2 mg/kg/Tantalum 2.13 mg/kg/Erbium 2.3 mg/kg/Europium 2.4 mg/kg/Sodium 2.49 %/Sodium oxide 3.36 %/Hafnium 3.6 mg/kg/Dysprosium 4.8 mg/kg/Magnesium 4.8 %/Lead 5.7 mg/kg/Thorium 6 mg/kg/Calcium 7.05 %/Samarium 7.8 mg/kg/Magnesium oxide 7.96 %/Iron 8.6 %/Aluminium 8.84 %/Rubidium 9.1 mg/kg/Calcium oxide 9.87 %/Praseodymium 12 mg/kg/Total Iron expressed as Fe2O3 12.3 %/Aluminium oxide 16.7 %/Gallium 20 mg/kg/Silicon 21.8 %/Yttrium 24.4 mg/kg/Scandium 27.1 mg/kg/Niobium 42 mg/kg/Silicon dioxide 46.6 %/Neodymium 47 mg/kg/Cobalt 50 mg/kg/Lanthanum 53.5 mg/kg/Copper 55 mg/kg/Chromium 86 mg/kg/Nickel 91 mg/kg/Zinc 102 mg/kg/Cerium 104 mg/kg/Zirconium 158 mg/kg/Vanadium 247 mg/kg/Barium 836 mg/kg/Strontium 978 mg/kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.