Learn More
Invitrogen™ MERS Coronavirus ORF4a Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119580
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, we recommend adding 50% glycerol, then aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Middle East respiratory syndrome-related coronavirus (MERS), or EMC/2012, is a species of coronavirus which infects humans, bats, and camels. The infecting virus is an enveloped, positive-sense, single-stranded RNA virus which enters its host cell by binding to the DPP4 receptor. The absence of all four Middle East respiratory syndrome CoV (MERS-CoV) accessory ORFs (deletion of ORF3, -4a, -4b, and -5 [dORF3-5]) has major implications for viral replication and pathogenesis.
Specifications
| MERS Coronavirus ORF4a | |
| Polyclonal | |
| Unconjugated | |
| MERS; Middle East Respiratory Syndrome | |
| Full length MERS-CoV ORF4a recombinant protein (MERS-CoV (strain United Kingdom/H123990006/2012)). | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability. | |
| Liquid |
| Western Blot | |
| 1.74 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.