missing translation for 'onlineSavingsMsg'
Learn More

MES monohydrate, free acid, ≥99%, MP Biomedicals™

Product Code. 11446581 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
250 g
Unit Size:
100g
10g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446581 10 g 10g
11456581 100 g 100g
11466581 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11446581 Supplier MP Biomedicals Supplier No. 0219483510

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A buffer using MES free acid can be prepared by titrating the free acid with sodium hydroxide to the desired pH (pKa ± 1). Alternatively, volumes of equimolar MES free acid and sodium or potassium MES can be mixed to attain the desired pH

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4432-31-9
Molecular Formula C6H13NO4S
Molecular Weight (g/mol) 195.233
MDL Number MFCD00149409
InChI Key SXGZJKUKBWWHRA-UHFFFAOYSA-N
Synonym 2-[N-morpholino]ethanesulfonic acid
PubChem CID 78165
ChEBI CHEBI:39005
IUPAC Name 2-morpholin-4-ylethanesulfonic acid
SMILES C1COCCN1CCS(=O)(=O)O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 10 g
Assay Percent Range ≥99%
Formula Weight 213.26
Percent Purity 99.6%
Chemical Name or Material MES monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.