missing translation for 'onlineSavingsMsg'
Learn More

Melford MES Sodium Salt

Product Code. 30018472
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018472 1 kg 1kg
30018473 500 g 500g
30018471 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018472 Supplier Melford Supplier No. M220501000.0
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MES Sodium Salt, also known as 2-(N-Morpholino)ethanesulfonic acid sodium salt, is a zwitterionic buffer commonly used in biological and biochemical research.

MES Sodium Salt, also known as 2-(N-Morpholino)ethanesulfonic acid sodium salt, is a zwitterionic buffer commonly used in biological and biochemical research.

In the life science field, MES Sodium Salt is primarily utilized as a buffering agent to maintain a stable pH in various experimental procedures. It is particularly effective in the pH range of 5.5 to 6.7, making it suitable for applications requiring slightly acidic to neutral pH conditions.

MES Sodium Salt is commonly used in protein purification, enzyme assays, and electrophoresis techniques where a specific pH range needs to be maintained. Its buffering capacity and stability within the desired pH range make it valuable in these applications.

One of the significant advantages of MES Sodium Salt is its compatibility with biological systems and enzymes. It exhibits minimal interference with enzymatic activity and other biological processes, ensuring reliable and accurate results.

Specifications

Chemical Name or Material MES Sodium Salt
CAS 71119-23-8
Quantity 1 kg
Molecular Formula C6H12NNaO4S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.