missing translation for 'onlineSavingsMsg'
Learn More

MES Sodium Salt, MP Biomedicals™

Product Code. 11492722 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492722 10 g 10g
11402732 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11492722 Supplier MP Biomedicals Supplier No. 0215245410
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 71119-23-8
Molecular Formula C6H12NNaO4S
Molecular Weight (g/mol) 217.215
MDL Number MFCD00065473
InChI Key IRHWMYKYLWNHTL-UHFFFAOYSA-M
Synonym mes sodium salt, sodium 2-morpholinoethanesulfonate, 2-n-morpholino ethanesulfonic acid sodium salt, 4-morpholineethanesulfonic acid, sodium salt, sodium 4-morpholin-1-ylethylsulphonate, sodium 2-4-morpholinyl ethanesulfonate, mes-na, 4-morpholineethanesulfonic acid sodium salt, sodium 4-morpholineethanesulfonate, 4-morpholineethanesulfonic acid, sodium salt 1:1
PubChem CID 23673676
ChEBI CHEBI:62955
IUPAC Name sodium;2-morpholin-4-ylethanesulfonate
SMILES C1COCCN1CCS(=O)(=O)[O-].[Na+]

Specifications

Color White
Assay Percent Range 1.003
Quantity 10 g
Formula Weight 217.2
Grade Not Available
Physical Form Powder
Chemical Name or Material MES
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.