missing translation for 'onlineSavingsMsg'
Learn More

MES Sodium Salt, TRC

Product Code. 30476228
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30476228 100 g 100g
30469287 25 g 25g
30471189 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30476228 Supplier Toronto Research Chemicals Supplier No. TRCM218775100G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71119-23-8
Molecular Formula C6H12NNaO4S
Molecular Weight (g/mol) 217.22
Synonym 2-(N-Morpholino)ethanesulfonic Acid Sodium Salt, 4-Morpholineethanesulfonic Acid, Sodium Salt (9CI), 2-(N-Morpholino)ethanesulfonate Sodium
IUPAC Name sodium;2-morpholin-4-ylethanesulfonate
SMILES [Na+].[O-]S(=O)(=O)CCN1CCOCC1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C6H13NO4S.Na/c8-12(9,10)6-3-7-1-4-11-5-2-7;/h1-6H2,(H,8,9,10);/q;+1/p-1
Formula Weight 217.0385
Chemical Name or Material MES Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.