missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MESDC2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10958-1-AP-150UL
This item is not returnable.
View return policy
Description
MESDC2 chaperone specifically assisting the folding of beta-propeller/EGF modules within the family of low-density lipoprotein receptors (LDLRs). Acts as a modulator of the Wnt pathway through chaperoning the coreceptors of the canonical Wnt pathway, LRP5 and LRP6, to the plasma membrane. Essential for specification of embryonic polarity and mesoderm induction. Plays an essential role in neuromuscular junction (NMJ) formation by promoting cell-surface expression of LRP4 (By similarity). May regulate phagocytosis of apoptotic retinal pigment epithelium (RPE) cells (By similarity).Specifications
| MESDC2 | |
| Polyclonal | |
| Unconjugated | |
| MESD | |
| BOCA, KIAA0081, LDLR chaperone MESD, MESD, MESDC2, Mesoderm development protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23184, 67943 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14696, Q9ERE7 | |
| Mesd | |
| MESDC2 Fusion Protein Ag1403 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction