missing translation for 'onlineSavingsMsg'
Learn More

Meta Fluoxetine Hydrochloride, TRC

Product Code. 30135429
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135429 25 mg 25mg
30134480 50 mg 50mg
30012344 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135429 Supplier Toronto Research Chemicals Supplier No. TRCM22582025MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79088-29-2
Molecular Formula C17 H18 F3 N O . Cl H
Molecular Weight (g/mol) 345.79
Synonym (3RS)-N-Methyl-3-phenyl-3-[3-(trifluoromethyl)phenoxy]propan-1-amine Hydrochloride, Fluoxetine Hydrochloride Imp. C (EP) as Hydrochloride
IUPAC Name N-methyl-3-phenyl-3-[3-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride
SMILES Cl.CNCCC(Oc1cccc(c1)C(F)(F)F)c2ccccc2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C17H18F3NO.ClH/c1-21-11-10-16(13-6-3-2-4-7-13)22-15-9-5-8-14(12-15)17(18,19)20;/h2-9,12,16,21H,10-11H2,1H3;1H
Formula Weight 345.1107
Percent Purity >95%
Chemical Name or Material (3RS)-N-Methyl-3-phenyl-3-[3-(trifluoromethyl)phenoxy]propan-1-amine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.