missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrachlorophenol in Isopropanol 100μg/mL, Fisher Chemical™

Product Code. 12933014 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12933014 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12933014 Supplier Fisher Chemical Supplier No. P872580

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

Packaging Ampoule
CAS 935-95-5
Physical Form Liquid
Molecular Formula C6H2Cl4O
Concentration 100 μg/mL
InChI Key KEWNKZNZRIAIAK-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES C1=C(C(=C(C(=C1Cl)Cl)O)Cl)Cl
IUPAC Name 2,3,5,6-tetrachlorophenol
Solvent or Matrix Isopropanol (2-propanol)
Molecular Weight (g/mol) 231.881
PubChem CID 13636
Quantity 5 mL
ChEBI CHEBI:52048
Chemical Name or Material 2,3,5,6-Tetrachlorophenol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.