missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2.3-Dibromopropionic acid in Methyl-tert.butylether 1000μg/mL, Fisher Chemical™
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule
Specifications
Concentration | 1000μg/mL |
CAS | 600-05-5 |
Packaging | Ampoule |
Quantity | 1 mL |
Type | Organic Single-component Standard Solution |
Molecular Formula | C3H4Br2O2 |
MDL Number | MFCD00004212 |
InChI Key | ZMYAKSMZTVWUJB-UHFFFAOYNA-N |
SMILES | OC(=O)C(Br)CBr |
IUPAC Name | 2,3-dibromopropanoic acid |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'