missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromophenol in DCM 10μg/mL, Fisher Chemical™

Product Code. 12920254 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12920254 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12920254 Supplier Fisher Chemical Supplier No. P855250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Packaging Ampoule
CAS 118-79-6
Physical Form Liquid
Molecular Formula C6H3Br3O
Concentration 10 μg/mL
MDL Number MFCD00002150
InChI Key BSWWXRFVMJHFBN-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES OC1=C(Br)C=C(Br)C=C1Br
IUPAC Name 2,4,6-tribromophenol
Solvent or Matrix Dichloromethane
Molecular Weight (g/mol) 330.80
PubChem CID 1483
Quantity 10 mL
ChEBI CHEBI:47696
Chemical Name or Material 2,4,6-Tribromophenol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.