missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromophenol in Ethanol 1000μg/mL, Fisher Chemical™

Product Code. 12936153 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12936153 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12936153 Supplier Fisher Chemical Supplier No. F131341

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 608-33-3
Molecular Formula C6H4Br2O
Concentration 1000 μg/mL
MDL Number MFCD00002152
InChI Key SSIZLKDLDKIHEV-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES OC1=C(Br)C=CC=C1Br
IUPAC Name 2,6-dibromophenol
Solvent or Matrix Ethanol
Molecular Weight (g/mol) 251.91
PubChem CID 11847
Quantity 1 mL
ChEBI CHEBI:19391
Chemical Name or Material 2,6-Dibromophenol
Show More Show Less
compliance-icons
Product Identifier
  • standard solution 2,6-dibromophenol 1000µg/ml in ethanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, 2,6-dibromophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.