missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenol in Methanol 2000μg/mL, Fisher Chemical™

Product Code. 12996093 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12996093 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12996093 Supplier Fisher Chemical Supplier No. F116591

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 367-12-4
Molecular Formula C6H5FO
Concentration 2000 μg/mL
MDL Number MFCD00002155
InChI Key HFHFGHLXUCOHLN-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES OC1=CC=CC=C1F
IUPAC Name 2-fluorophenol
Solvent or Matrix Methanol
Molecular Weight (g/mol) 112.10
PubChem CID 9707
Quantity 1 mL
Chemical Name or Material 2-Fluorophenol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.