Learn More
4-tert-Butylphenol in Ethanol 1000μg/mL, Fisher Chemical™
Product Code. 12963114 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12963114 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12963114 Supplier Fisher Chemical Supplier No. P873660
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Packaging Ampoule
CAS 98-54-4
Physical Form Liquid
Molecular Formula C10H14O
Concentration 1000 μg/mL
InChI Key QHPQWRBYOIRBIT-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES CC(C)(C)C1=CC=C(C=C1)O
IUPAC Name 4-tert-butylphenol
Solvent or Matrix Ethanol
Molecular Weight (g/mol) 150.221
PubChem CID 7393
Quantity 1 mL
ChEBI CHEBI:34444
Chemical Name or Material 4-tert-Butylphenol