missing translation for 'onlineSavingsMsg'
Learn More

Metabolite Chlorthal-dimethyl in Methanol 100μg/mL, Fisher Chemical™

Product Code. 12952614 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12952614 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12952614 Supplier Fisher Chemical Supplier No. P815320

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Packaging Ampoule(s)
CAS 1861-32-1
Molecular Formula C10H6Cl4O4
Concentration 100 μg/mL
MDL Number MFCD00014902
InChI Key NPOJQCVWMSKXDN-UHFFFAOYSA-N
SMILES COC(=O)C1=C(Cl)C(Cl)=C(C(=O)OC)C(Cl)=C1Cl
IUPAC Name 1,4-dimethyl 2,3,5,6-tetrachlorobenzene-1,4-dicarboxylate
Solvent or Matrix Methanol
Molecular Weight (g/mol) 331.95
PubChem CID 2943
Quantity 1 mL
ChEBI CHEBI:34664
Chemical Name or Material Chlorthal-dimethyl
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.