missing translation for 'onlineSavingsMsg'
Learn More

Metabolite Chrysene in Cyclohexane 100μg/mL, Fisher Chemical™

Product Code. 12912544 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12912544 10 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12912544 Supplier Fisher Chemical Supplier No. P813060

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Packaging Ampoule(s)
CAS 218-01-9
Molecular Formula C18H12
Concentration 10 μg/mL
UN Number 1145
InChI Key WDECIBYCCFPHNR-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=CC3=C2C=CC4=CC=CC=C43
IUPAC Name chrysene
Solvent or Matrix Cyclohexane
Molecular Weight (g/mol) 228.294
PubChem CID 9171
Quantity 10 mL
ChEBI CHEBI:51687
Chemical Name or Material Chrysene
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Standard solution Chrysene 100ug/ml in cyclohexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Aspiration toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P321-Specific treatment (see on this label).
  • P331-Do NOT induce vomiting.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
  • P391-Collect spillage.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.