missing translation for 'onlineSavingsMsg'
Learn More

Metabolite Decafluorobiphenyl in DCM 100μg/mL, Fisher Chemical™

Product Code. 12952654 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12952654 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12952654 Supplier Fisher Chemical Supplier No. P816120

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Packaging Ampoule(s)
CAS 434-90-2
Molecular Formula C12F10
Concentration 100 μg/mL
InChI Key ONUFSRWQCKNVSL-UHFFFAOYSA-N
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)C2=C(C(=C(C(=C2F)F)F)F)F
IUPAC Name 1,2,3,4,5-pentafluoro-6-(2,3,4,5,6-pentafluorophenyl)benzene
Solvent or Matrix Dichloromethane
Molecular Weight (g/mol) 334.116
PubChem CID 67949
Quantity 1 mL
Chemical Name or Material Decafluorobiphenyl
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.