missing translation for 'onlineSavingsMsg'
Learn More

Dibenz[a,h]acridine in Methanol 100μg/mL, Fisher Chemical™

Product Code. 12905884 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12905884 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12905884 Supplier Fisher Chemical Supplier No. P875280

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule(s)
CAS 226-36-8
Molecular Formula C21H13N
Concentration 100 μg/mL
InChI Key JNCSIWAONQTVCF-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=CC3=C2C=C4C=CC5=CC=CC=C5C4=N3
Solvent or Matrix Methanol
Molecular Weight (g/mol) 279.342
PubChem CID 9183
Quantity 1 mL
ChEBI CHEBI:82310
Chemical Name or Material Dibenz[a,h]acridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.