missing translation for 'onlineSavingsMsg'
Learn More

Metabolite Flurochloridone in Cyclohexane 100μg/mL, Fisher Chemical™

Product Code. 12926793 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12926793 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12926793 Supplier Fisher Chemical Supplier No. P826680

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

Packaging Ampoule(s)
CAS 61213-25-0
Molecular Formula C12H10Cl2F3NO
Concentration 100 μg/mL
MDL Number MFCD00144317
UN Number 1145
InChI Key OQZCSNDVOWYALR-UHFFFAOYNA-N
SMILES FC(F)(F)C1=CC=CC(=C1)N1CC(CCl)C(Cl)C1=O
IUPAC Name 3-chloro-4-(chloromethyl)-1-[3-(trifluoromethyl)phenyl]pyrrolidin-2-one
Solvent or Matrix Cyclohexane
Molecular Weight (g/mol) 312.11
PubChem CID 91677
Quantity 1 mL
ChEBI CHEBI:5131
Chemical Name or Material Flurochloridone
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Standard solution Flurochloridone 100ug/ml in cyclohexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Aspiration toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P321-Specific treatment (see on this label).
  • P331-Do NOT induce vomiting.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
  • P391-Collect spillage.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.