missing translation for 'onlineSavingsMsg'
Learn More

Metabolite Oxadixyl in Methanol 100μg/mL, Fisher Chemical™

Product Code. 12992744 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 12992744

Brand: Fisher Chemical P835960

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

Packaging Ampoule(s)
CAS 77732-09-3
Molecular Formula C14H18N2O4
Concentration 100mu;g/mL
MDL Number MFCD00145052
InChI Key UWVQIROCRJWDKL-UHFFFAOYSA-N
SMILES COCC(=O)N(N1CCOC1=O)C1=C(C)C=CC=C1C
IUPAC Name N-(2,6-dimethylphenyl)-2-methoxy-N-(2-oxo-1,3-oxazolidin-3-yl)acetamide
Solvent or Matrix Iso-octane
Molecular Weight (g/mol) 278.31
PubChem CID 53735
Quantity 1 mL
ChEBI CHEBI:81937
Chemical Name or Material Oxadixyl
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.