Learn More
Metabolite Phenanthrene D10 in Methyl-tert.butylether 100μg/mL, Fisher Chemical™
Product Code. 12906383 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12906383 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12906383 Supplier Fisher Chemical Supplier No. P847830
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Specifications
Packaging Ampoule
CAS 1517-22-2
Molecular Formula C14H10
Concentration 100 μg/mL
InChI Key YNPNZTXNASCQKK-LHNTUAQVSA-N
SMILES C1=CC=C2C(=C1)C=CC3=CC=CC=C32
IUPAC Name 1,2,3,4,5,6,7,8,9,10-decadeuteriophenanthrene
Solvent or Matrix Methyl-tert.butylether
Molecular Weight (g/mol) 188.295
PubChem CID 519070
Quantity 1 mL
Chemical Name or Material Phenanthrene D10