missing translation for 'onlineSavingsMsg'
Learn More

Procymidone in Methanol 100μg/mL, Fisher Chemical™

Product Code. 12906443 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12906443 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12906443 Supplier Fisher Chemical Supplier No. P849000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Packaging Ampoule
CAS 32809-16-8
Physical Form Liquid
Molecular Formula C13H11Cl2NO2
Concentration 100 μg/mL
InChI Key QXJKBPAVAHBARF-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES CC12CC1(C(=O)N(C2=O)C3=CC(=CC(=C3)Cl)Cl)C
IUPAC Name 3-(3,5-dichlorophenyl)-1,5-dimethyl-3-azabicyclo[3.1.0]hexane-2,4-dione
Solvent or Matrix Methanol
Molecular Weight (g/mol) 284.136
PubChem CID 36242
Quantity 1 mL
Chemical Name or Material Procymidone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.