missing translation for 'onlineSavingsMsg'
Learn More

Triazophos in Methanol 100μg/mL, Fisher Chemical™

Product Code. 12970244 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12970244 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12970244 Supplier Fisher Chemical Supplier No. P855100
  • ONLINE EXCLUSIVE PRICE
    284.57€
    197.73€ / 1mL
    Save 86.84€
    30% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 24017-47-8
Physical Form Liquid
Molecular Formula C12H16N3O3PS
Concentration 100 μg/mL
UN Number 1230
InChI Key AMFGTOFWMRQMEM-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES CCOP(=S)(OCC)OC1=NN(C=N1)C2=CC=CC=C2
IUPAC Name diethoxy-[(1-phenyl-1,2,4-triazol-3-yl)oxy]-sulfanylidene-$l^{5}-phosphane
Solvent or Matrix Methanol
Molecular Weight (g/mol) 313.312
PubChem CID 32184
Quantity 1 mL
ChEBI CHEBI:38963
Chemical Name or Material Triazophos
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Standard solution Triazophos,O,O-diethyl-O-1-phenyl-1H-1,2,4-triazol-3-yl phosphorothioate 100ug/ml in methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H370-Causes damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P361+P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.