Learn More
Metal Additives Standard, MA5: Ba, Ca, Mg, P, Zn @ 900 μg/g in Hydrocarbon Oil, VHG™
Description
Metal Additives Standard, MA5: Ba, Ca, Mg, P, Zn @ 900 μg/g in Hydrocarbon Oil
Specifications
Specifications
| Chemical Name or Material | Phosphorus/Magnesium/Calcium/Barium/Zinc |
| CAS | 7723-14-0, 7439-95-4, 7440-70-2, 7440-39-3, 7440-66-6 |
| Methods | ICP/XRF/RDE |
| Analyte or Component Names | Phosphorus/Magnesium/Calcium/Barium/Zinc |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | 75 cSt Hydrocarbon Oil |
| Quantity | 50 g |
| Concentration or Composition (by Analyte or Components) | Phosphorus 896.9 μg/g/Magnesium 897.4 μg/g/Calcium 897.9 μg/g/Barium 898.5 μg/g/Zinc 898.7 μg/g |